Linoleic acid ≥99%, Ultrapure
Catalog # 0660-10G
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:10 g
- Seq:3236
Specifications
Specification Test Results
- Purity:≥99 %
Specification Test Results
About this item
:
:280.45 g/mol
Merck Index:13,05527
Boiling Pt:360 °C (1013 hPa)
Melting Pt:–5 °C
Density:0.9 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-470-9
CAS Number:60-33-3
Formula:C₁₈H₃₂O₂
MDL Number:MFCD00064241