Ethyl sorbate, Sigma-Aldrich®
Catalog # SIAL177687-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:140.18 g/mol
Boiling Pt:195.4 °C (1013 hPa)
Density:0.9405 g/cm³ (15.3 °C)
:M-Clarity™ quality level = MQ100
EINECS:219-258-2
CAS Number:2396-84-1
Formula:C₈H₁₂O₂
MDL Number:MFCD00009296