Order Entry
Export
Orders LinkContactUsLinkComponent
Ethyl linoleate

Ethyl linoleate

Glass bottle for solids, 1 g
Catalog # ROTH3198.1
Supplier:   Roth Carl
Ethyl linoleate
Catalog # ROTH3198.1
Supplier:  Roth Carl
CAS Number:  544-35-4
Ethyl linoleate

Glass bottle for solids, 1 g

CAS Number:  544-35-4
Price
41,50 €
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  9Z,12Z-Octadecadienoic acid ethyl ester, Linoleic acid ethyl ester, cis,cis-9,12-Octadecadienoic acid ethyl ester, (9Z,12Z)-Ethyl octadeca-9,12-dienoate
  • CAS Number:544-35-4
  • Formula:C₂₀H₃₆O₂
  • MDL Number:MFCD00009535
  • Molecular Weight:308.5 g/mol
  • Storage Temperature:Freezer
  • EINECS:208-868-4
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications