


Hexachloro-1,3-butadiene
Catalog # EHERC14170000
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,25 g
- Seq:3029
Specifications
About this item
:
:260.76 g/mol
Merck Index:13,04697
Boiling Pt:210 °C (1013 hPa)
Melting Pt:–21 °C
Density:1.6825 g/cm³ (20 °C)
Flash Pt:90 °C
EINECS:201-765-5
UN:2279
ADR:6.1,III
:GHS06
:GHS05
:GHS08
Storage Temperature:Refrigerator
CAS Number:87-68-3
Formula:C₄Cl₆
MDL Number:MFCD00000836