Hexachloro-1,3-butadiene ≥97%
Glass bottle for solids, 50 g
Supplier: Thermo Fisher Scientific
Hexachloro-1,3-butadiene ≥97%
Catalog # A17316.18
Supplier: Thermo Fisher Scientific
CAS Number: 87-68-3
Jump to:
Product Details & Documents
Purity: 97% CAS No.: 87-68-3 Molecular Formula: CCl2=CClCCl=CCl2 Formula Weight: 260.76 Refractive Index: 1.5550 Odour: Mild Boiling Point: 215° Beilstein Reference: 1766570 Merck Reference: 14,4678 UN No.: 2279 MDL No.: MFCD00000836
- CAS Number:87-68-3
- Formula:C₄Cl₆
- MDL Number:MFCD00000836
- Molecular Weight:260.76 g/mol
- Merck Index:14,4678
- Boiling Pt:215 °C
- Melting Pt:–21 °C
- Density:1.680
- Flash Pt:90 °C
- Storage Temperature:Ambient
- EINECS:201-765-5
- UN:2279
- ADR:6.1,III
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:50 g



