Order Entry
Export
Orders LinkContactUsLinkComponent
Bathophenanthroline, Sigma-Aldrich®

Bathophenanthroline, Sigma-Aldrich®

Supplier:   Merck
Bathophenanthroline, Sigma-Aldrich®
Supplier:  Merck
Certificates
Bathophenanthroline, Sigma-Aldrich®

2 Options Available Below

Product Details & Documents
Synonyms:  4,7-Diphenyl-1,10-phenanthroline, (Bathophenanthroline)
  • CAS Number:1662-01-7
  • Formula:C₂₄H₁₆N₂
  • MDL Number:MFCD00004976
  • Molecular Weight:332.4 g/mol
  • Boiling Pt:508 °C (1013 hPa)
  • Melting Pt:215…220 °C
  •  :M-Clarity™ quality level = MQ200
  • Storage Temperature:Ambient
  • EINECS:216-767-1
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    500 mg
    Log in for availability
    Log in for availability
    1 * 500 mg
    1 * 500 mg - 109,00 €
    109,00 €
    1 * 500 mg
       109,00 €

  • Glass bottle for solids
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 181,00 €
    181,00 €
    1 * 1 g
       181,00 €