Bathophenanthroline ≥98%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 98+% CAS No.: 1662-01-7 Molecular Formula: C24H16N2 Formula Weight: 332.41 Melting Point: 218-222° Beilstein Reference: 261048
Synonyms: 4,7-Diphenyl-1,10-phenanthroline, (Bathophenanthroline)
- CAS Number:1662-01-7
- Formula:C₂₄H₁₆N₂
- MDL Number:MFCD00004976
- Molecular Weight:332.4 g/mol
- Boiling Pt:508 °C (1013 hPa)
- Melting Pt:218…222 °C
- Storage Temperature:Ambient
- EINECS:216-767-1
3 Options Available Below
Product Details & Documents
Purity: 98+% CAS No.: 1662-01-7 Molecular Formula: C24H16N2 Formula Weight: 332.41 Melting Point: 218-222° Beilstein Reference: 261048
Synonyms: 4,7-Diphenyl-1,10-phenanthroline, (Bathophenanthroline)
- CAS Number:1662-01-7
- Formula:C₂₄H₁₆N₂
- MDL Number:MFCD00004976
- Molecular Weight:332.4 g/mol
- Boiling Pt:508 °C (1013 hPa)
- Melting Pt:218…222 °C
- Storage Temperature:Ambient
- EINECS:216-767-1



