D-(+)-Biotin, Sigma-Aldrich®
Catalog # SIAL19606-1G-F
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:244.31 g/mol
Merck Index:13,01231
Melting Pt:232…233 °C
Density:1.41 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Storage Temperature:Refrigerator
EINECS:200-399-3
CAS Number:58-85-5
Formula:C₁₀H₁₆N₂O₃S
MDL Number:MFCD00005541