D-(+)-Biotin
Jump to:
About this item
Synonyms: Vitamin H, D-Biotin, Vitamin B8, 5-((3aS,4S,6aR)-2-Oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanoic acid, Vitamin B7
- CAS Number:58-85-5
- Formula:C₁₀H₁₆N₂O₃S
- MDL Number:MFCD00005541
- Molecular Weight:244.31 g/mol
- Merck Index:13,01231
- Melting Pt:232…233 °C
- Density:1.41 g/cm³ (20 °C)
- Storage Temperature:Refrigerator
- EINECS:200-399-3
2 Options Available Below
Synonyms: Vitamin H, D-Biotin, Vitamin B8, 5-((3aS,4S,6aR)-2-Oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanoic acid, Vitamin B7
- CAS Number:58-85-5
- Formula:C₁₀H₁₆N₂O₃S
- MDL Number:MFCD00005541
- Molecular Weight:244.31 g/mol
- Merck Index:13,01231
- Melting Pt:232…233 °C
- Density:1.41 g/cm³ (20 °C)
- Storage Temperature:Refrigerator
- EINECS:200-399-3
Product Family Options
Product Information
- Pack typePk



