D-(+)-Biotin
Glass bottle, 1 mL
Catalog # FITZ20-BR20
Supplier: FITZGERALD INDUSTRIES INTL
Jump to:
Product Details & Documents
Synonyms: Vitamin H, D-Biotin, Vitamin B8, 5-((3aS,4S,6aR)-2-Oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanoic acid, Vitamin B7
- CAS Number:58-85-5
- Formula:C₁₀H₁₆N₂O₃S
- MDL Number:MFCD00005541
- Molecular Weight:244.31 g/mol
- Merck Index:13,01231
- Melting Pt:232…233 °C
- Density:1.41 g/cm³ (20 °C)
- Storage Temperature:Refrigerator
- EINECS:200-399-3
Specifications
Specifications
- Pack type:Glass bottle
- Pk:1 mL



