(R)-(-)-2-Methylpiperazine, Sigma-Aldrich®
Catalog # SIAL397164-1G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:100.16 g/mol
Boiling Pt:153 °C (1013 hPa)
Melting Pt:89…91 °C
ADR:4.1,II
:M-Clarity™ quality level = MQ100
UN:1325
CAS Number:75336-86-6
Formula:C₅H₁₂N₂
MDL Number:MFCD00192317