L(-)-Tryptophan, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:73-22-3
:
Formula:C₁₁H₁₂N₂O₂
MDL Number:MFCD00064340
:204.23 g/mol
Merck Index:12,09929
Melting Pt:290 °C
Density:1.34 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-795-6