Order Entry
Export
Orders LinkContactUsLinkComponent
L(-)-Tryptophan

L(-)-Tryptophan

Supplier:   Roth Carl
L(-)-Tryptophan
Supplier:  Roth Carl
Certificates

3 Options Available Below

Product Details & Documents
Synonyms:  L-Tryptophan, Tryptophan, L-α-Amino-3-indolepropionic acid, (S)-2-Amino-3-(3-indolyl)propionic acid, H-Trp-OH
  • CAS Number:73-22-3
  • Formula:C₁₁H₁₂N₂O₂
  • MDL Number:MFCD00064340
  • Molecular Weight:204.23 g/mol
  • Merck Index:12,09929
  • Melting Pt:290 °C
  • Density:1.34 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-795-6
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    10 g
    Log in for availability
    Log in for availability
    1 * 10 g
    1 * 10 g - 18,60 €
    18,60 €
    1 * 10 g
       18,60 €

  • Glass bottle for solids
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 91,90 €
    91,90 €
    1 * 100 g
       91,90 €

  • Glass bottle for solids
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg - 732,00 €
    732,00 €
    1 * 1 kg
       732,00 €