L(+)-Histidine
Catalog # MOLEM47484029
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:155.16 g/mol
Merck Index:13,04741
Melting Pt:272…273 °C
Density:1.44 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-745-3
CAS Number:71-00-1
Formula:C₆H₉N₃O₂
MDL Number:MFCD00064315