L(+)-Leucine
Catalog # A1426.1000
Supplier: PanReac AppliChem
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 kg
- Seq:4024
Specifications
About this item
:
:131.17 g/mol
Merck Index:14,05451
Melting Pt:300 °C
Density:1.165 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-522-0
CAS Number:61-90-5
Formula:C₆H₁₃NO₂
MDL Number:MFCD00002617