L(+)-Valine, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:72-18-4
:
Formula:C₅H₁₁NO₂
MDL Number:MFCD00064220
:117.15 g/mol
Merck Index:13,09458
Boiling Pt:217 °C (1013 hPa)
Melting Pt:86 °C
Density:1.284 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-773-6