Order Entry
Export
Orders LinkContactUsLinkComponent
D-(-)-Sorbitol, Sigma-Aldrich®

D-(-)-Sorbitol, Sigma-Aldrich®

Supplier:   Merck
D-(-)-Sorbitol, Sigma-Aldrich®
Supplier:  Merck
Certificates
D-(-)-Sorbitol, Sigma-Aldrich®
D-(-)-Sorbitol, Sigma-Aldrich®

2 Options Available Below

Product Details & Documents
Synonyms:  (2R,3R,4R,5S)-Hexane-1,2,3,4,5,6-hexaol, (Sorbitol), D(-)-Glucitol, D-Sorbitol, Sorbitol
  • CAS Number:50-70-4
  • Formula:HOH₂C(CH(OH))₄CH₂OH
  • MDL Number:MFCD00004708
  • Molecular Weight:182.17 g/mol
  • Merck Index:13,08797
  • Boiling Pt:295 °C (1013 hPa)
  • Melting Pt:94…98 °C
  • Density:1.524 g/cm³ (20 °C)
  •  :M-Clarity™ quality level = MQ200
  • Flash Pt:>100 °C
  • Storage Temperature:Ambient
  • EINECS:200-061-5

Documents

Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 38,80 €
    38,80 €
    1 * 5 g
       38,80 €

  • Glass bottle for solids
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 65,50 €
    65,50 €
    1 * 100 g
       65,50 €