ESD safety shoes, clogs, Michel
Supplier: ELTEN
Certificates
About this item
Lightweight anti-static clogs made from smooth, tear resistant microfibre upper, an adjustable Velcro® heel strap, and a mono-density PU sole (2,5 mm, oil, fuel and abrasion resistant, heat resistant up to approx. 130 °C).
- Climate-regulating, breathable textile lining
- Full length inlay sole (C-Fit ESD)
- ESD soft fleece insole
- Shock-absorbent heel
- Protective steel toecap (200 J)
Colour: Black or white
Certifications: EN 61340-5-1 (ESD); EN ISO 20345 SB, SRC, A, E, FO.
Specifications
- Catalog No:
- ELTN72430/40
- ELTN72440/42
- ELTN72440/41
- ELTN72440/40
- ELTN72430/48
- ELTN72430/47
- ELTN72430/46
- ELTN72440/48
- ELTN72430/45
- ELTN72440/47
- ELTN72440/46
- ELTN72430/44
- ELTN72440/45
- ELTN72430/43
- ELTN72440/44
- ELTN72430/42
- ELTN72440/43
- ELTN72430/41
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001