D-(-)-Mannitol
Catalog # PROONE9283
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mL
- Seq:2275
Specifications
About this item
:
:182.17 g/mol
Merck Index:13,05769
Boiling Pt:290…295 °C (4 hPa)
Melting Pt:164…169 °C
Density:1.49 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-711-8
CAS Number:69-65-8
Formula:HOH₂C(CH(OH))₄CH₂OH
MDL Number:MFCD00064287