(S,S)-(+)-2,4-Pentanediol
Catalog # MOLE28444806-1G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:104.15 g/mol
Boiling Pt:111…113 °C (19 mmHg)
Melting Pt:45…48 °C
Storage Temperature:Ambient
CAS Number:72345-23-4
Formula:C₅H₁₂O₂
MDL Number:MFCD00064276