(S)-(+)-1,3-Butanediol
Catalog # MOLE10179508-1G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:90.12 g/mol
Boiling Pt:109 °C (1013 hPa)
Density:1 g/cm³ (23 °C)
EINECS:246-363-0
CAS Number:24621-61-2
Formula:C₄H₁₀O₂
MDL Number:MFCD00064278
Boiling Pt:109 °C (1013 hPa)