(S)-(+)-1,2-Propanediol, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:4254-15-3
:
Formula:C₃H₈O₂
MDL Number:MFCD00004539
:76.1 g/mol
Density:1.03 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
Storage Temperature:Ambient