Order Entry
Export
ContactUsLinkComponent
(S)-(+)-1,2-Propanediol
(S)-(+)-1,2-Propanediol
Catalog # MOLE43121408-1G
Supplier:  Molekula
CAS Number:  
(S)-(+)-1,2-Propanediol
Catalog # MOLE43121408-1G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :76.1 g/mol
Density:1.03 g/cm³ (20 °C)
Storage Temperature:Ambient
CAS Number:4254-15-3
Formula:C₃H₈O₂
MDL Number:MFCD00004539