Order Entry
Export
ContactUsLinkComponent
(R,R)-(-)-2,4-Pentanediol
(R,R)-(-)-2,4-Pentanediol
Catalog # MOLE63669768-1G
Supplier:  Molekula
CAS Number:  
(R,R)-(-)-2,4-Pentanediol
Catalog # MOLE63669768-1G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
Boiling Pt:207 °C (1013 hPa)
Melting Pt:50.5 °C
 :104.15 g/mol
Storage Temperature:Ambient
CAS Number:42075-32-1
Formula:C₅H₁₂O₂
MDL Number:MFCD00063893