Order Entry
Export
ContactUsLinkComponent
 
(2R,3R)-(-)-2,3-Butanediol 98+%, 99+% ee
 
undefined
(2R,3R)-(-)-2,3-Butanediol 98+%, 99+% ee

 

  • Appearance:
    Clear colorless Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4309 to 1.4329 (20°C, 589 nm)
  • GC:
    ≥98.0 %
  • Enantiomeric excess:
    ≥99.0 % (GC)
  • Specific optical rotation:
    -12.5° to -13.5° (20°C, 589 nm) (neat)