Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
(R)-(-)-1,3-Butanediol
(R)-(-)-1,3-Butanediol, Sigma-Aldrich®
(R)-(-)-1,3-Butanediol, Sigma-Aldrich®
(R)-(-)-1,3-Butanediol, Sigma-Aldrich®
SIAL237612-1G
:
Merck
:
(R)-(-)-1,3-Butanediol, Sigma-Aldrich®
SIAL237612-1G
:
Merck
:
Pack type:
Glass bottle for solids
Pk:
1 g
Seq:
3047
:
:
90.12 g/mol
:
1 g/cm³ (20 °C)
:
M-Clarity™ quality level = MQ100
:
228-532-0
:
6290-03-5
:
C₄H₁₀O₂
:
MFCD00064277