Order Entry
Export
Orders LinkContactUsLinkComponent
(R)-(-)-1,3-Butanediol

(R)-(-)-1,3-Butanediol

Glass bottle for solids, 1 g
Catalog # MOLE63144414-1G
Supplier:   Molekula
(R)-(-)-1,3-Butanediol
Catalog # MOLE63144414-1G
Supplier:  Molekula
CAS Number:  6290-03-5
(R)-(-)-1,3-Butanediol

Glass bottle for solids, 1 g

CAS Number:  6290-03-5
Price
Inquire For Price
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  (R)-(-)-1,3-Dihydroxy butane, (R)-(-)-1,3-Butylene glykol, (R)-Butane-1,3-diol
  • CAS Number:6290-03-5
  • Formula:C₄H₁₀O₂
  • MDL Number:MFCD00064277
  • Molecular Weight:90.12 g/mol
  • Density:1 g/cm³ (20 °C)
  • EINECS:228-532-0
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications