Order Entry
Export
Orders LinkContactUsLinkComponent
 

(R)-(-)-1,2-Propanediol, Sigma-Aldrich®

Supplier:   Merck
(R)-(-)-1,2-Propanediol, Sigma-Aldrich®
Supplier:  Merck
Certificates
(R)-(-)-1,2-Propanediol, Sigma-Aldrich®

2 Options Available Below

Product Details & Documents
Synonyms:  (R)-(-)-1,2-Dihydroxy propane, (R)-(-)-1,2-Propylene glykol, (R)-Propane-1,2-diol
  • CAS Number:4254-14-2
  • Formula:C₃H₈O₂
  • MDL Number:MFCD00066248
  • Molecular Weight:76.1 g/mol
  •  :M-Clarity™ quality level = MQ100
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 40,00 €
    40,00 €
    1 * 5 g
       40,00 €

  • Glass bottle for solids
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 128,00 €
    128,00 €
    1 * 25 g
       128,00 €