L(+)-Potassium sodium tartrate tetrahydrate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: Potassium sodium tartrate tetrahydrate, Potassium sodium (2R,3R)-2,3-dihydroxysuccinate tetrahydrate, Potassium sodium L(+)-tartrate tetrahydrate, Sodium potassium tartrate, (R,R)-(+)-Tartaric acid potassium sodium salt tetrahydrate, (R,R)-(+)-Potassium sodium tartrate tetrahydrate, Rochelle salt, L(+)-Tartaric acid potassium sodium salt tetrahydrate, (+)-Potassium sodium L-tartrate tetrahydrate
CAS Number:6381-59-5
:
Formula:KOOC(CHOH)₂COONa·4H₂O
MDL Number:MFCD00150989
:282.22 g/mol
Merck Index:13,07755
Melting Pt:70…80 °C
Density:∼1.767 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:206-156-8
Specifications
- Environmentally Preferable: