L(+)-Potassium hydrogen tartrate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:868-14-4
:
Formula:KOOC(CHOH)₂COOH
MDL Number:MFCD00065392
:188.18 g/mol
Merck Index:13,07697
Melting Pt:250 °C
Density:1.943 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
EINECS:212-769-1