L(+)-Potassium hydrogen tartrate ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:868-14-4
:
Formula:KOOC(CHOH)₂COOH
MDL Number:MFCD00065392
:188.18 g/mol
Merck Index:14,7615
Melting Pt:∼ 267 °C (dec.)
Density:1.954
Storage Temperature:Ambient
EINECS:212-769-1