L(+)-Potassium hydrogen tartrate 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:868-14-4
:
Formula:KOOC(CHOH)₂COOH
MDL Number:MFCD00065392
:188.18 g/mol
Merck Index:13,07697
Melting Pt:250 °C
Density:1.95 g/cm³
Storage Temperature:Ambient
EINECS:212-769-1
Specification Test Results
- Appearance:Colorless to white Powder or crystals
- Infrared spectrum:Conforms
- Loss on drying:≤0.5 % (105°C)
- Specific optical rotation:+8.0° to +9.2° (20°C, 589 nm) (c=10, HCl 1M and H2O) (on dried substance)
- Titration with NaOH:98.5 to 101.5 % (On dried substance)
- Chloride (Cl):≤500 ppm
- Sulfate (SO4):≤500 ppm