ContactUsLinkComponent
L(+)-Tartaric acid
  MOLEM82341396
 :  Molekula
 :  
undefined
L(+)-Tartaric acid
  MOLEM82341396
 :  Molekula
 :  
  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024
  • Environmentally Preferable:
 : L-Tartaric Acid, (2R,3R)-(+)-Tartaric acid, Tartaric Acid, (R,R)-(+)-2,3-Dihydroxysuccinic acid, (R,R)-(+)-Tartaric acid, (2R,3R)-2,3-Dihydroxysuccinic acid
 :
 :150.09 g/mol
 :399
 :150 °C (closed cup)
 :201-766-0
 :01-2119537204-47
 :GHS07
 :Ambient
 :87-69-4
 :C₄H₆O₆
 :MFCD00064207
 :168…170 °C
 :1.759 g/cm³ (20 °C)
 :13,09158