D-(+)-Malic acid
Supplier: Roth Carl
Certificates
About this item
Synonyms: (R)-(+)-Hydroxysuccinic acid, D(+)-Hydroxy-1,4-butanedioic acid, (R)-(+)-Hydroxy-1,4-butanedioic acid, D(+)-Malic acid, D(+)-Hydroxysuccinic acid, (R)-2-Hydroxysuccinic acid
CAS Number:636-61-3
:
Formula:C₄H₆O₅
MDL Number:MFCD00004245
Boiling Pt:322
Molecular Weight:134.09 g/mol
Melting Pt:101…104 °C
Merck Index:13,05307
EINECS:211-262-2
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable: