Organic reference standard, 3,3'-Dichlorobiphenyl (PCB No. 11) 10 µg/ml in isooctane, Dr Ehrenstorfer
Catalog # EHERL20001100IO
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:2050-67-1
- Description:3,3'-Dichlorobiphenyl (PCB No. 11) 10 µg/ml in isooctane
- Formula:C₁₂H₈Cl₂
- Description:3,3'-Dichlorobiphenyl (PCB No. 11)