Organic reference standard, 3,3',4-Trichlorobiphenyl (PCB No. 35) 10 µg/ml in isooctane, Dr Ehrenstorfer
Catalog # EHERL20003500IO
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:37680-69-6
- Description:3,3',4-Trichlorobiphenyl (PCB No. 35) 10 µg/ml in isooctane
- Formula:C₁₂H₇Cl₃
- Description:3,3',4-Trichlorobiphenyl (PCB No. 35)