Organic reference standard, 3,3',4,4',5-Pentachlorobiphenyl (PCB No. 126) 10 µg/ml in isooctane, Dr Ehrenstorfer
Catalog # EHERL20012600IO
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:57465-28-8
- Description:3,3',4,4',5-Pentachlorobiphenyl (PCB No. 126) 10 µg/ml in isooctane
- Formula:C₁₂H₅Cl₅
- Description:3,3',4,4',5-Pentachlorobiphenyl (PCB No. 126)