Calcium-D gluconate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:299-28-5
:
Formula:HOH₂C(CH(OH))₄COOCa
MDL Number:MFCD00064209
:430.38 g/mol
Merck Index:13,01671
Melting Pt:178 °C
:M-Clarity™ quality level = MQ200
EINECS:206-075-8