Order Entry
Export
ContactUsLinkComponent
Calcium-D gluconate
Calcium-D gluconate
Catalog # MOLE51659757-100G
Supplier:  Molekula
CAS Number:  
Calcium-D gluconate
Catalog # MOLE51659757-100G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :430.38 g/mol
Merck Index:13,01671
Melting Pt:178 °C
EINECS:206-075-8
CAS Number:299-28-5
Formula:HOH₂C(CH(OH))₄COOCa
MDL Number:MFCD00064209