Sodium gluconate for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: D-Gluconic acid sodium salt, Sodium (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate, Gluconic acid, sodium salt, Sodium D-gluconate
CAS Number:527-07-1
:
Formula:HOH₂C(CH(OH))₄COONa
MDL Number:MFCD00064210
Molecular Weight:218.14 g/mol
Melting Pt:170…175 °C
Merck Index:12,08766
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:208-407-7
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Solubility:590 g/l (25 °C)
- Melting Point:170 - 175 °C
- Molar Mass:218.14 g/mol
- pH:7.0 - 8.0 (100 g/l, H2O, 20 °C)
- Ignition Point:200 °C
Specification Test Results
Related Information
Certificates
Search by Lot NumberMERC8.22058.2500_TDS_EN ProductPage_822058
Product specification
msds_822058