Sodium gluconate 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:527-07-1
:
Formula:HOH₂C(CH(OH))₄COONa
MDL Number:MFCD00064210
Merck Index:12,08766
Melting Pt:206…209 °C (dec.)
:218.14 g/mol
Storage Temperature:Ambient
EINECS:208-407-7
Specification Test Results
- Appearance:White to light beige Crystalline powder
- Infrared spectrum:Conforms
- pH:6.5 to 7.5 (10 % solution)
- Titration with HClO4:≥97.5 %
- Chloride (Cl):≤100 ppm
- Lead (Pb):≤10 ppm
- Reducing substances:≤0.5 %