L(+)-Lithium lactate 99%, pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:27848-80-2
:
Formula:C₃H₅LiO₃
MDL Number:MFCD00065512
:96.01 g/mol
Boiling Pt:200 °C
Melting Pt:>75 °C
Storage Temperature:Ambient
EINECS:248-692-5
Specification Test Results
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Specific optical rotation:-13° to -16° (20°C, 589 nm) (c=1, H2O)
- Titration with HClO4:≥98.5 %