Organic reference standard, 2,2',4,5,5'-Pentachlorobiphenyl (PCB No. 101) 10 µg/ml in isooctane, Dr Ehrenstorfer
EHERL20010100IO
:
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:37680-73-2
- Description:2,2',4,5,5'-Pentachlorobiphenyl (PCB No. 101) 10 µg/ml in isooctane
- Formula:C₁₂H₅Cl₅
- Description:2,2',4,5,5'-Pentachlorobiphenyl (PCB No. 101)