Order Entry
Export
Orders LinkContactUsLinkComponent
L(+)-Cysteine

L(+)-Cysteine

Glass bottle for solids, 1 kg
Catalog # A1425.1000
Irritant
Supplier:   PanReac AppliChem
L(+)-Cysteine
Catalog # A1425.1000
Supplier:  PanReac AppliChem
CAS Number:  52-90-4
L(+)-Cysteine

Glass bottle for solids, 1 kg

CAS Number:  52-90-4
Price
2 070,00 €
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  H-Cys-OH, L-(+)-Cysteine, (R)-2-Amino-3-mercaptopropanoic acid
  • CAS Number:52-90-4
  • Formula:HSCH₂CH(NH₂)CO₂H
  • MDL Number:MFCD00064306
  • Molecular Weight:121.16 g/mol
  • Merck Index:13,02810
  • Boiling Pt:254 °C (1013 hPa)
  • Melting Pt:115 °C
  • Density:1.523 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-158-2
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg

Specifications