Organic reference standard, 2,2',3,3',4,4',6-Heptachlorobiphenyl (PCB No. 171) 10 µg/ml in isooctane, Dr Ehrenstorfer
Catalog # EHERL20017100IO
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:52663-71-5
- Description:2,2',3,3',4,4',6-Heptachlorobiphenyl (PCB No. 171) 10 µg/ml in isooctane
- Formula:C₁₂H₃Cl₇
- Description:2,2',3,3',4,4',6-Heptachlorobiphenyl (PCB No. 171)