Order Entry
Export
Orders LinkContactUsLinkComponent
L(+)-Glutamine, Sigma-Aldrich®

L(+)-Glutamine, Sigma-Aldrich®

Supplier:   Merck
L(+)-Glutamine, Sigma-Aldrich®
Supplier:  Merck
Certificates
L(+)-Glutamine, Sigma-Aldrich®
L(+)-Glutamine, Sigma-Aldrich®

3 Options Available Below

Product Details & Documents
Synonyms:  Gln, L-Glutamine, H-Gln-OH, (S)-(+)-Glutamine, (S)-2,5-Diamino-5-oxopentanoic acid, Levoglutamide, Q
  • CAS Number:56-85-9
  • Formula:C₅H₁₀N₂O₃
  • MDL Number:MFCD00008044
  • Molecular Weight:146.15 g/mol
  • Merck Index:13,04484
  • Boiling Pt:355 °C (1013 hPa)
  • Melting Pt:185…186 °C
  • Density:1.5326 g/cm³ (130 °C)
  • Storage Temperature:Ambient
  • EINECS:200-292-1
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 60,70 €
    60,70 €
    1 * 25 g
       60,70 €

  • Glass bottle for solids
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 193,00 €
    193,00 €
    1 * 100 g
       193,00 €

  • Glass bottle for solids
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 609,00 €
    609,00 €
    1 * 500 g
       609,00 €