Order Entry
Export
ContactUsLinkComponent
L(+)-Glutamine
L(+)-Glutamine
Catalog # EHERC14034450
Supplier:  EHRENSTORFER
CAS Number:  
undefined
L(+)-Glutamine
Catalog # EHERC14034450
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,1 g
  • Seq:
    3020

Specifications

About this item

 :
 :146.15 g/mol
Merck Index:13,04484
Boiling Pt:355 °C (1013 hPa)
Melting Pt:185…186 °C
Density:1.5326 g/cm³ (130 °C)
Storage Temperature:Ambient
EINECS:200-292-1
CAS Number:56-85-9
Formula:C₅H₁₀N₂O₃
MDL Number:MFCD00008044