Organic reference standard, Toluene 100 µg/ml in methanol, Dr Ehrenstorfer
Catalog # EHERXA17594000ME
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:108-88-3
- Description:Toluene 100 µg/ml in methanol
- Formula:C₇H₈
- Description:Toluene