Pesticide reference standard, 4-Chlorotoluene 100 µg/ml in methanol, Dr Ehrenstorfer
EHERXA11522000ME
:
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:106-43-4
- Description:4-Chlorotoluene 100 µg/ml in methanol
- Formula:C₇H₇Cl
- Description:4-Chlorotoluene