Organic reference standard, 1-Bromo-4-fluorobenzene 2000 µg/ml in methanol, Dr Ehrenstorfer
Catalog # EHERYA10731000ME
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:460-00-4
- Description:1-Bromo-4-fluorobenzene 2,000 µg/ml in methanol
- Formula:C₆H₄BrF
- Description:4-Bromofluorobenzene